Anti-DRAK2 Rabbit Polyclonal Antibody [clone: I286]
Catalog # BWRLBS9183
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:DRAK2
- Clonality:Polyclonal
- Clone:I286
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 261-310 of Human DRAK2.
DRAK2 (I286) pAb detects endogenous levels of DRAK2 protein.
Type: Primary
Antigen: DRAK2
Clonality: Polyclonal
Clone: I286
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat