Anti-Urotensin II Rabbit Polyclonal Antibody
BWRLBS7278
- Antibody type:Primary
- Antigen symbol:Urotensin II
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Recombinant full length Human Urotensin II.
Urotensin II pAb detects endogenous levels of Urotensin II protein.
Type: Primary
Antigen: Urotensin II
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human