Anti-p-PR Rabbit Polyclonal Antibody [clone: S190]
Catalog # BWRLBS4158
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-PR
- Clonality:Polyclonal
- Clone:S190
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human PR around the phosphorylation site of Serine 190.
p-Progesterone Receptor (S190) pAb detects endogenous levels of Progesterone Receptor protein only when phosphorylated at Ser190.
Type: Primary
Antigen: p-PR
Clonality: Polyclonal
Clone: S190
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human