Anti-p-ABL1 Rabbit Polyclonal Antibody [clone: Y412]
BWRLBS5002
- Antibody type:Primary
- Antigen symbol:p-ABL1
- Clonality:Polyclonal
- Clone:Y412
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human ABL1 around the phosphorylation site of Tyrosine 412.
p-ABL1 (Y412) pAb detects endogenous levels of ABL1 protein when phosphorylated at Tyr412.
Type: Primary
Antigen: p-ABL1
Clonality: Polyclonal
Clone: Y412
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat