Order Entry
Poland
ContactUsLinkComponent
Anti-p-PP2A-alpha Rabbit Polyclonal Antibody [clone: Y307]
Anti-p-PP2A-alpha Rabbit Polyclonal Antibody [clone: Y307]
Catalog # BWRLBS4867
Anti-p-PP2A-alpha Rabbit Polyclonal Antibody [clone: Y307]
Catalog # BWRLBS4867

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-PP2A-alpha
  • Clonality:
    Polyclonal
  • Clone:
    Y307
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human PP2A-alpha around the phosphorylation site of Tyrosine 307.

p-PP2A-? (Y307) pAb detects endogenous levels of PP2A-? protein only when phosphorylated at Tyr307.

Type: Primary
Antigen: p-PP2A-alpha
Clonality: Polyclonal
Clone: Y307
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat