Anti-p-PKC delta Rabbit Polyclonal Antibody [clone: Y52]
BWRLBS4861
- Antibody type:Primary
- Antigen symbol:p-PKC delta
- Clonality:Polyclonal
- Clone:Y52
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human PKC delta around the phosphorylation site of Tyrosine 52.
p-PKC ? (Y52) pAb detects endogenous levels of PKC ? protein only when phosphorylated at Tyr52.
Type: Primary
Antigen: p-PKC delta
Clonality: Polyclonal
Clone: Y52
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat