Anti-MT-ATP8 Rabbit Polyclonal Antibody
Catalog # BWRLBS60106
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MT-ATP8
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human MT-ATP8.
MT-ATP8 pAb detects endogenous levels of MT-ATP8 protein.
Type: Primary
Antigen: MT-ATP8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat