Anti-p-NOS3 Rabbit Polyclonal Antibody [clone: S1177]
Catalog # BWRLBS4848
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-NOS3
- Clonality:Polyclonal
- Clone:S1177
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human NOS3 around the phosphorylation site of Serine 1177.
p-NOS3 (S1177) pAb detects endogenous levels of NOS3 protein when phosphorylated at Ser1177.
Type: Primary
Antigen: p-NOS3
Clonality: Polyclonal
Clone: S1177
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat