Anti-p-p27 Kip1 Rabbit Polyclonal Antibody [clone: T187]
Catalog # BWRLBS4838
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-p27 Kip1
- Clonality:Polyclonal
- Clone:T187
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human p27 Kip1 around the phosphorylation site of Threonine 187.
p-p27 Kip1 (T187) pAb detects endogenous levels of p27 Kip1 protein when phosphorylated at Thr187.
Type: Primary
Antigen: p-p27 Kip1
Clonality: Polyclonal
Clone: T187
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat