Anti-p-Dok-2 Rabbit Polyclonal Antibody [clone: Y299]
BWRLBS4786
- Antibody type:Primary
- Antigen symbol:p-Dok-2
- Clonality:Polyclonal
- Clone:Y299
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human Dok-2 around the phosphorylation site of Tyrosine 299.
p-Dok-2 (Y299) pAb detects endogenous levels of Dok-2 protein when phosphorylated at Tyr299.
Type: Primary
Antigen: p-Dok-2
Clonality: Polyclonal
Clone: Y299
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse