Anti-p-Cortactin Rabbit Polyclonal Antibody [clone: Y421]
Catalog # BWRLBS4778
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Cortactin
- Clonality:Polyclonal
- Clone:Y421
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 360-418 of Human TFDP1.
p-Cortactin (Y421) pAb detects endogenous levels of Cortactin protein only when phosphorylated at Tyr421.
Type: Primary
Antigen: p-Cortactin
Clonality: Polyclonal
Clone: Y421
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat