Anti-p-ERK 1/2 Rabbit Polyclonal Antibody [clone: T202]
Catalog # BWRLBS4759
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-ERK 1/2
- Clonality:Polyclonal
- Clone:T202
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human ERK1/2 around the phosphorylation site of Threonine 202.
p-ERK 1/2 (T202) pAb detects endogenous levels of ERK 1/2 protein only when phosphorylated at Thr202.
Type: Primary
Antigen: p-ERK 1/2
Clonality: Polyclonal
Clone: T202
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat