Anti-p-Cdk1/Cdc2 Rabbit Polyclonal Antibody [clone: Y15]
Catalog # BWRLBS4758
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Cdk1/Cdc2
- Clonality:Polyclonal
- Clone:Y15
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Cdk1/Cdc2 around the phosphorylation site of Serine 795.
p-Cdk1/Cdc2 (Y15) pAb detects endogenous levels of Cdk1/Cdc2 protein only when phosphorylated at Tyr15.
Type: Primary
Antigen: p-Cdk1/Cdc2
Clonality: Polyclonal
Clone: Y15
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat