Anti-p-FoxO4 Rabbit Polyclonal Antibody [clone: T451]
Catalog # BWRLBS4687
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-FoxO4
- Clonality:Polyclonal
- Clone:T451
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human FoxO4 around the phosphorylation site of Threonine 451.
p-FoxO4 (T451) pAb detects endogenous levels of FoxO4 protein only when phosphorylated at Thr198.
Type: Primary
Antigen: p-FoxO4
Clonality: Polyclonal
Clone: T451
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse