Anti-p-FAK Rabbit Polyclonal Antibody [clone: Y576]
Catalog # BWRLBS4684
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-FAK
- Clonality:Polyclonal
- Clone:Y576
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human FAK around the phosphorylation site of Serine 576.
p-FAK (Y576) pAb detects endogenous levels of FAK protein only when phosphorylated at Tyr576.
Type: Primary
Antigen: p-FAK
Clonality: Polyclonal
Clone: Y576
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat