Anti-p-CK18 Rabbit Polyclonal Antibody [clone: S52]
BWRLBS4676
- Antibody type:Primary
- Antigen symbol:p-CK18
- Clonality:Polyclonal
- Clone:S52
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human Cytokeratin 18 around the phosphorylation site of Serine 53.
p-Cytokeratin 18 (S52) pAb detects endogenous levels of Cytokeratin 18 protein only when phosphorylated at Ser52. (removal of the initiator methionine)
Type: Primary
Antigen: p-CK18
Clonality: Polyclonal
Clone: S52
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat