Anti-p-Crystallin-alphaB Rabbit Polyclonal Antibody [clone: S19]
Catalog # BWRLBS4672
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Crystallin-alphaB
- Clonality:Polyclonal
- Clone:S19
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Crystallin-alphaB around the phosphorylation site of Serine 19.
p-Crystallin-?B (S19) pAb detects endogenous levels of Crystallin-?B protein only when phosphorylated at Ser19.
Type: Primary
Antigen: p-Crystallin-alphaB
Clonality: Polyclonal
Clone: S19
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat