Anti-p-Chk1 Rabbit Polyclonal Antibody [clone: S286]
Catalog # BWRLBS4670
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Chk1
- Clonality:Polyclonal
- Clone:S286
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Chk1 around the phosphorylation site of Serine 286.
p-Chk1 (S286) pAb detects endogenous levels of Chk1 protein when phosphorylated at Ser286.
Type: Primary
Antigen: p-Chk1
Clonality: Polyclonal
Clone: S286
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat