Anti-p-NPM Rabbit Polyclonal Antibody [clone: T199]
Catalog # BWRLBS4655
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-NPM
- Clonality:Polyclonal
- Clone:T199
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human NPM around the phosphorylation site of Threonine 199.
p-NPM (T199) pAb detects endogenous levels of NPM protein only when phosphorylated at Thr199.
Type: Primary
Antigen: p-NPM
Clonality: Polyclonal
Clone: T199
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat