Anti-p-FAK Rabbit Polyclonal Antibody [clone: Y407]
Catalog # BWRLBS4406
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-FAK
- Clonality:Polyclonal
- Clone:Y407
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human FAK around the phosphorylation site of Serine 407.
p-FAK (Y407) pAb detects endogenous levels of FAK only when phosphorylated at Tyr407.
Type: Primary
Antigen: p-FAK
Clonality: Polyclonal
Clone: Y407
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat