Anti-CASP9 Rabbit Polyclonal Antibody [clone: A149]
Catalog # BWRLBS1830
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CASP9
- Clonality:Polyclonal
- Clone:A149
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 120-170 of Human Caspase 9.
Caspase 9 (A149) pAb detects endogenous levels of Caspase 9 protein.
Type: Primary
Antigen: CASP9
Clonality: Polyclonal
Clone: A149
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human