Anti-FBP3 Rabbit Polyclonal Antibody [clone: R241]
Catalog # BWRLBS3297
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:FBP3
- Clonality:Polyclonal
- Clone:R241
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 210-254 of Human FBP3.
FBP3 (R241) pAb detects endogenous levels of FBP3 protein.
Type: Primary
Antigen: FBP3
Clonality: Polyclonal
Clone: R241
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat