Anti-PR Rabbit Polyclonal Antibody [clone: A394]
BWRLBS1766
- Antibody type:Primary
- Antigen symbol:PR
- Clonality:Polyclonal
- Clone:A394
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 361-410 of Human PR.
PR (A394) pAb detects endogenous levels of PR protein.
Type: Primary
Antigen: PR
Clonality: Polyclonal
Clone: A394
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human