Anti-CLC-4 Rabbit Polyclonal Antibody [clone: E254]
Catalog # BWRLBS3103
Dostawca: Bioworld Technology
Dane techniczne
- Antibody type:Primary
- Antigen symbol:CLC-4
- Clonality:Polyclonal
- Clone:E254
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Dane techniczne
Informacje o tym produkcie
Synthetic peptide, corresponding to amino acids 240-280 of Human CLC-4.
CLC-4 (E254) pAb detects endogenous levels of CLC-4 protein.
Type: Primary
Antigen: CLC-4
Clonality: Polyclonal
Clone: E254
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat