Anti-RPS4Y1 Rabbit Polyclonal Antibody [clone: K254]
Catalog # BWRLBS3044
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:RPS4Y1
- Clonality:Polyclonal
- Clone:K254
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 225-270 of Human Ribosomal Protein S4Y1.
Ribosomal Protein S4Y1 (K254) pAb detects endogenous levels of Ribosomal Protein S4Y1 protein.
Type: Primary
Antigen: RPS4Y1
Clonality: Polyclonal
Clone: K254
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human