Anti-RPL10 Rabbit Polyclonal Antibody [clone: F34]
Catalog # BWRLBS2549
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:RPL10
- Clonality:Polyclonal
- Clone:F34
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human RPL10.
RPL10 (F34) pAb detects endogenous levels of RPL10 protein.
Type: Primary
Antigen: RPL10
Clonality: Polyclonal
Clone: F34
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat