Anti-Histone H2A.X Rabbit Polyclonal Antibody [clone: S130]
Catalog # BWRLBS2984
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Histone H2A.X
- Clonality:Polyclonal
- Clone:S130
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 100-150 of Human Histone H2A.X.
Histone H2A.X (S130) pAb detects endogenous levels of Histone H2A.X protein.
Type: Primary
Antigen: Histone H2A.X
Clonality: Polyclonal
Clone: S130
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat