Anti-VPS72 Rabbit Polyclonal Antibody [clone: Q149]
Catalog # BWRLBS2487
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:VPS72
- Clonality:Polyclonal
- Clone:Q149
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 116-168 of Human VPS72.
VPS72 (Q149) pAb detects endogenous levels of VPS72 protein.
Type: Primary
Antigen: VPS72
Clonality: Polyclonal
Clone: Q149
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse