Anti-p68 RNA Helicase Rabbit Polyclonal Antibody [clone: N589]
Catalog # BWRLBS1630
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p68 RNA Helicase
- Clonality:Polyclonal
- Clone:N589
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 550-600 of Human p68 RNA Helicase.
p68 RNA Helicase (N589) pAb detects endogenous levels of p68 RNA Helicase protein.
Type: Primary
Antigen: p68 RNA Helicase
Clonality: Polyclonal
Clone: N589
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat