Anti-DDX3 Rabbit Polyclonal Antibody [clone: L318]
BWRLBS1629
- Antibody type:Primary
- Antigen symbol:DDX3
- Clonality:Polyclonal
- Clone:L318
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 291-340 of Human DDX3.
DDX3 (L318) pAb detects endogenous levels of DDX3 protein.
Type: Primary
Antigen: DDX3
Clonality: Polyclonal
Clone: L318
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat