Anti-TK Rabbit Polyclonal Antibody [clone: P7]
Catalog # BWRLBS1601
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:TK
- Clonality:Polyclonal
- Clone:P7
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human TK.
TK (P7) pAb detects endogenous levels of TK protein.
Type: Primary
Antigen: TK
Clonality: Polyclonal
Clone: P7
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat