Anti-Arrestin-C Rabbit Polyclonal Antibody [clone: E374]
BWRLBS2394
- Antibody type:Primary
- Antigen symbol:Arrestin-C
- Clonality:Polyclonal
- Clone:E374
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 350-400 of Human Arrestin-C.
Arrestin-C (E374) pAb detects endogenous levels of Arrestin-C protein.
Type: Primary
Antigen: Arrestin-C
Clonality: Polyclonal
Clone: E374
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human