Anti-MRCKbeta Rabbit Polyclonal Antibody [clone: F1665]
Catalog # BWRLBS2382
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:MRCK beta
- Clonality:Polyclonal
- Clone:F1665
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 1630-1680 of Human MRCKbeta.
MRCK? (F1665) pAb detects endogenous levels of MRCK? protein.
Type: Primary
Antigen: MRCKbeta
Clonality: Polyclonal
Clone: F1665
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat