Order Entry
Poland
ContactUsLinkComponent
Anti-MKP-4 Rabbit Polyclonal Antibody [clone: A174]
Anti-MKP-4 Rabbit Polyclonal Antibody [clone: A174]
  BWRLBS2348
Anti-MKP-4 Rabbit Polyclonal Antibody [clone: A174]
  BWRLBS2348

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MKP-4
  • Clonality:
    Polyclonal
  • Clone:
    A174
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 140-184 of Human MKP-4.

MKP-4 (A174) pAb detects endogenous levels of MKP-4 protein.

Type: Primary
Antigen: MKP-4
Clonality: Polyclonal
Clone: A174
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat