Anti-Neuropilin-2 Rabbit Polyclonal Antibody [clone: M373]
Catalog # BWRLBS3687
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Neuropilin-2
- Clonality:Polyclonal
- Clone:M373
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 341-390 of Human Neuropilin-2.
Neuropilin-2 (M373) pAb detects endogenous levels of Neuropilin-2 protein.
Type: Primary
Antigen: Neuropilin-2
Clonality: Polyclonal
Clone: M373
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat