Anti-BMP-7 Rabbit Polyclonal Antibody [clone: E173]
Catalog # BWRLBS3674
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:BMP-7
- Clonality:Polyclonal
- Clone:E173
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 140-190 of Human BMP-7.
BMP-7 (E173) pAb detects endogenous levels of BMP-7 protein.
Type: Primary
Antigen: BMP-7
Clonality: Polyclonal
Clone: E173
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat