Anti-Cathepsin D Rabbit Polyclonal Antibody [clone: K345]
BWRLBS3673
- Antibody type:Primary
- Antigen symbol:Cathepsin D
- Clonality:Polyclonal
- Clone:K345
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 311-360 of Human Cathepsin D.
Cathepsin D (K345) pAb detects endogenous levels of Cathepsin D protein.
Type: Primary
Antigen: Cathepsin D
Clonality: Polyclonal
Clone: K345
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat