Anti-14-3-3 sigma Rabbit Polyclonal Antibody [clone: E66]
Catalog # BWRLBS2178
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 sigma
- Clonality:Polyclonal
- Clone:E66
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 40-90 of Human 14-3-3 sigma.
14-3-3 ? (E66) pAb detects endogenous levels of 14-3-3 ? protein
Type: Primary
Antigen: 14-3-3 sigma
Clonality: Polyclonal
Clone: E66
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse