Anti-Cortactin Rabbit Polyclonal Antibody [clone: Y460]
BWRLBS3642
- Antibody type:Primary
- Antigen symbol:Cortactin
- Clonality:Polyclonal
- Clone:Y460
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 427-481 of Human Cortactin.
Cortactin (Y460) pAb detects endogenous levels of Cortactin protein.
Type: Primary
Antigen: Cortactin
Clonality: Polyclonal
Clone: Y460
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat