Anti-Connexin 43 Rabbit Polyclonal Antibody [clone: R361]
Catalog # BWRLBS3640
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Connexin 43
- Clonality:Polyclonal
- Clone:R361
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 330-380 of Human Connexin 43.
Connexin 43 (R361) pAb detects endogenous levels of Connexin 43 protein.
Type: Primary
Antigen: Connexin 43
Clonality: Polyclonal
Clone: R361
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat