Anti-Op18 Rabbit Polyclonal Antibody [clone: V31]
BWRLBS3616
- Antibody type:Primary
- Antigen symbol:Op18
- Clonality:Polyclonal
- Clone:V31
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminus of Human Op18.
Op18 (V31) pAb detects endogenous levels of Op18 protein.
Type: Primary
Antigen: Op18
Clonality: Polyclonal
Clone: V31
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat