Anti-p-C/EBP-beta Rabbit Polyclonal Antibody [clone: T235/188]
BWRLBS4029
- Antibody type:Primary
- Antigen symbol:p-C/EBP- beta
- Clonality:Polyclonal
- Clone:T235/188
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human C/EBP-beta around the phosphorylation site of Threonine 235.
p-C/EBP-? (T235/188) pAb detects endogenous levels of human LAP only when phosphorylated at Thr235, mouse and rat LAP only when phosphorylated at Thr188, and LIP only when phosphorylated at Thr37.
Type: Primary
Antigen: p-C/EBP-beta
Clonality: Polyclonal
Clone: T235/188
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat