Anti-p-Bcl-2 Rabbit Polyclonal Antibody [clone: S70]
BWRLBS4023
- Antibody type:Primary
- Antigen symbol:p-Bcl-2
- Clonality:Polyclonal
- Clone:S70
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human Bcl-2 around the phosphorylation site of Serine 70.
p-Bcl-2 (S70) pAb detects endogenous levels of Bcl-2 protein only when phosphorylated at Ser70.
Type: Primary
Antigen: p-Bcl-2
Clonality: Polyclonal
Clone: S70
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat