Anti-p-ASK1 Rabbit Polyclonal Antibody [clone: S966]
BWRLBS4014
- Antibody type:Primary
- Antigen symbol:p-ASK1
- Clonality:Polyclonal
- Clone:S966
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human ASK1 around the phosphorylation site of Serine 966.
p-ASK1 (S966) pAb detects endogenous levels of ASK1protein only when phosphorylated atSer966.
Type: Primary
Antigen: p-ASK1
Clonality: Polyclonal
Clone: S966
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat