Anti-CD172b Rabbit Polyclonal Antibody [clone: V270]
Catalog # BWRLBS3947
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CD172b
- Clonality:Polyclonal
- Clone:V270
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 251-300 of Human CD172b.
CD172b (V270) pAb detects endogenous levels of CD172b protein.
Type: Primary
Antigen: CD172b
Clonality: Polyclonal
Clone: V270
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human