Anti-CASP12 Rabbit Polyclonal Antibody [clone: P99]
BWRLBS3542
- Antibody type:Primary
- Antigen symbol:CASP12
- Clonality:Polyclonal
- Clone:P99
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 65-123 of Human Caspase 12.
Caspase12 (P99) pAb detects endogenous levels of Caspase12 protein.
Type: Primary
Antigen: CASP12
Clonality: Polyclonal
Clone: P99
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat