Anti-CD13 Rabbit Polyclonal Antibody [clone: Q917]
Catalog # BWRLBS3531
Supplier: Bioworld Technology
Specifications
- Typ przeciwciała:Primary
- Symbol antygenu:CD13
- Klonalność:Poliklonalne
- Klonuj:Q917
- Gospodarz:Rabbit
- ImmunoChemia:Yes
- Reaktywność:Human,Rat,Mouse
- Western blot:Yes
- Rozmiar:100 µg
- Ilość:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 880-930 of Human CD13.
CD13 (Q917) pAb detects endogenous levels of CD13 protein.
Type: Primary
Antigen: CD13
Clonality: Polyclonal
Clone: Q917
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat