Anti-RRS1 Rabbit Polyclonal Antibody [clone: R286]
BWRLBS3932
- Antibody type:Primary
- Antigen symbol:RRS1
- Clonality:Polyclonal
- Clone:R286
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 255-307 of Human RRS1.
RRS1 (R286) pAb detects endogenous levels of RRS1 protein.
Type: Primary
Antigen: RRS1
Clonality: Polyclonal
Clone: R286
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat