Anti-AR-alpha2C Rabbit Polyclonal Antibody [clone: R369]
Catalog # BWRLBS1945
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:AR-alpha2C
- Clonality:Polyclonal
- Clone:R369
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 340-390 of Human AR-alpha2C.
AR ?2C (R369) pAb detects endogenous levels of AR ?2C protein.
Type: Primary
Antigen: AR-alpha2C
Clonality: Polyclonal
Clone: R369
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat