Anti-Separase Rabbit Polyclonal Antibody [clone: A795]
BWRLAP0374
- Antibody type:Primary
- Antigen symbol:Separase
- Clonality:Polyclonal
- Clone:A795
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 763-824 of Human Separase.
Separase (A795) pAb detects endogenous levels of Separase protein.
Type: Primary
Antigen: Separase
Clonality: Polyclonal
Clone: A795
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat